Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006372.1 | NP_001006372.1 | 6 - 7 | 534 | 75 | |
XM_015275875.2 | XP_015131361.1 | 6 - 7 | 507 | 75 | ||
GenBank mRNA | AJ720064.1 | - | 6 - 7 | 534 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |