Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012559.1 | NP_001012577.1 | 11 - 12 | 1209 | 136 | |
XM_015281547.2 | XP_015137033.1 | 13 - 14 | 2040 | 136 | ||
XM_015281548.2 | XP_015137034.1 | 12 - 13 | 2037 | 136 | ||
XM_015281549.2 | XP_015137035.1 | 12 - 13 | 1169 | 136 | ||
GenBank mRNA | AJ851567.1 | - | 11 - 12 | 1209 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |