Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012566.1 | NP_001012584.1 | 8 - 9 | 917 | 79 | |
GenBank mRNA | AJ851766.1 | - | 8 - 9 | 917 | 79 | |
CR352609.1 | - | 7 - 8 | 767 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |