Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012569.1 | NP_001012587.1 | 4 - 5 | 800 | 91 | |
GenBank mRNA | AJ851416.1 | - | 4 - 5 | 800 | 91 | |
BX936265.1 | - | 4 - 5 | 783 | 91 | ||
BX950282.1 | - | 4 - 5 | 506 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |