Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012569.1 | NP_001012587.1 | 6 - 7 | 975 | 90 | |
GenBank mRNA | AJ851416.1 | - | 6 - 7 | 975 | 90 | |
BX950282.1 | - | 6 - 7 | 681 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |