Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012579.1 | NP_001012597.1 | 4 - 5 | 696 | 115 | |
XM_015276240.2 | XP_015131726.1 | 4 - 5 | 866 | 115 | ||
GenBank mRNA | AY899804.1 | - | 4 - 5 | 696 | 115 | |
BX930336.2 | - | 4 - 5 | 656 | 115 | ||
BX932595.2 | - | 4 - 5 | 942 | 115 | ||
BX936134.2 | - | 4 - 5 | 929 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |