Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012594.2 | NP_001012612.1 | 5 - 6 | 632 | 99 | |
XM_015289644.2 | XP_015145130.1 | 5 - 6 | 603 | 99 | ||
GenBank mRNA | AJ851797.1 | - | 5 - 6 | 634 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |