Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012690.1 | NP_001012708.1 | 5 - 6 | 578 | 111 | |
XM_015292095.2 | XP_015147581.1 | 5 - 6 | 903 | 111 | ||
GenBank mRNA | AJ851484.1 | - | 5 - 6 | 578 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |