Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278083.1 | NP_001265012.1 | 7 - 8 | 734 | 133 | |
XM_025148612.1 | XP_025004380.1 | 5 - 6 | 592 | 133 | ||
GenBank mRNA | AJ851408.1 | - | 5 - 6 | 886 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |