Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012699.1 | NP_001012717.1 | 6 - 7 | 820 | 151 | |
GenBank mRNA | AJ851448.1 | - | 6 - 7 | 820 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |