Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012797.1 | NP_001012815.1 | 9 - 10 | 1279 | 64 | |
GenBank mRNA | AJ851610.1 | - | 9 - 10 | 1279 | 64 | |
CR338808.1 | - | 1 - 2 | 142 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |