Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012844.2 | NP_001012862.2 | 11 - 12 | 1193 | 97 | |
NM_001305191.1 | NP_001292120.1 | 12 - 13 | 1246 | 97 | ||
NM_001305192.1 | NP_001292121.1 | 12 - 13 | 1222 | 97 | ||
NM_001305193.1 | NP_001292122.1 | 10 - 11 | 1135 | 97 | ||
NM_001305194.1 | NP_001292123.1 | 11 - 12 | 1217 | 97 | ||
XM_025143970.1 | XP_024999738.1 | 13 - 14 | 3240 | 97 | ||
XM_025143971.1 | XP_024999739.1 | 13 - 14 | 3216 | 97 | ||
GenBank mRNA | AJ720237.1 | - | 11 - 12 | 1193 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |