Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012861.1 | NP_001012879.1 | 5 - 6 | 604 | 114 | |
XM_015275856.2 | XP_015131342.1 | 5 - 6 | 1090 | 114 | ||
XM_015275857.2 | XP_015131343.1 | 5 - 6 | 671 | 114 | ||
XM_015275858.2 | XP_015131344.1 | 4 - 5 | 913 | 114 | ||
XM_015275859.2 | XP_015131345.1 | 4 - 5 | 889 | 114 | ||
GenBank mRNA | AJ719502.1 | - | 5 - 6 | 604 | 114 | |
CR338760.1 | - | 5 - 6 | 579 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |