Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012868.2 | NP_001012886.1 | 654 | 83 | ||
GenBank mRNA | AJ720026.1 | - | 654 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus |