Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012887.2 | NP_001012905.2 | 2 - 3 | 642 | 88 | |
GenBank mRNA | AJ720987.1 | - | 2 - 3 | 642 | 88 | |
CR387179.1 | - | 1 - 2 | 119 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |