Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012892.1 | NP_001012910.1 | 6 - 7 | 677 | 110 | |
XM_025149734.1 | XP_025005502.1 | 6 - 7 | 110 | |||
GenBank mRNA | AJ719851.1 | - | 6 - 7 | 677 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |