Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012892.1 | NP_001012910.1 | 7 - 8 | 830 | 88 | |
XM_025149734.1 | XP_025005502.1 | 7 - 8 | 951 | 88 | ||
GenBank mRNA | AJ719851.1 | - | 7 - 8 | 830 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |