Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC40A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012913.1 | NP_001012931.1 | 2 - 3 | 333 | 113 | |
GenBank mRNA | AJ851647.1 | - | 2 - 3 | 333 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |