Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012945.1 | NP_001012963.1 | 7 - 8 | 1088 | 85 | |
GenBank mRNA | AJ720331.1 | - | 7 - 8 | 1088 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |