Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012997.1 | NP_001013015.1 | 3 - 4 | 361 | 85 | |
GenBank mRNA | BX931585.2 | - | 3 - 4 | 359 | 85 | |
BX950478.1 | - | 3 - 4 | 361 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |