Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014968.1 | NP_001014968.1 | 3 - 4 | 799 | 104 | |
XM_015293967.2 | XP_015149453.1 | 4 - 5 | 1109 | 104 | ||
XM_015293968.2 | XP_015149454.1 | 4 - 5 | 1031 | 104 | ||
XM_025154805.1 | XP_025010573.1 | 5 - 6 | 4044 | 104 | ||
GenBank mRNA | AF143240.1 | - | 3 - 4 | 686 | 104 | |
AY953144.1 | - | 3 - 4 | 799 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |