Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017413.1 | NP_001017413.1 | 2 - 3 | 957 | 60 | |
GenBank mRNA | AY672618.1 | - | 2 - 3 | 957 | 60 | |
BX950436.2 | - | 2 - 3 | 795 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |