Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024694.1 | NP_001019865.1 | 5 - 6 | 712 | 69 | |
XM_025153846.1 | XP_025009614.1 | 4 - 5 | 69 | |||
GenBank mRNA | BX929548.2 | - | 1 - 2 | 176 | 69 | |
BX935536.2 | - | 5 - 6 | 712 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |