Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029980.2 | NP_001025151.1 | 10 - 11 | 953 | 61 | |
XM_015290542.2 | XP_015146028.1 | 9 - 10 | 887 | 61 | ||
XM_015290544.2 | XP_015146030.1 | 8 - 9 | 896 | 61 | ||
XM_025149080.1 | XP_025004848.1 | 7 - 8 | 777 | 61 | ||
XM_025149081.1 | XP_025004849.1 | 9 - 10 | 942 | 61 | ||
XM_025149082.1 | XP_025004850.1 | 10 - 11 | 872 | 61 | ||
GenBank mRNA | CR353501.1 | - | 10 - 11 | 953 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |