Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2W1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642141.3 | XP_003642189.2 | 2 - 3 | 456 | 69 | |
XM_015294328.2 | XP_015149814.1 | 2 - 3 | 1309 | 69 | ||
XM_025155214.1 | XP_025010982.1 | 2 - 3 | 263 | 69 | ||
GenBank mRNA | AJ720136.1 | - | 457 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |