Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSFL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030859.1 | NP_001026030.1 | 1 - 2 | 110 | 136 | |
GenBank mRNA | AJ720274.1 | - | 1 - 2 | 110 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |