Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSFL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030859.1 | NP_001026030.1 | 8 - 9 | 951 | 72 | |
XM_015296393.2 | XP_015151879.1 | 8 - 9 | 1181 | 72 | ||
GenBank mRNA | AJ720274.1 | - | 8 - 9 | 951 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |