Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030877.1 | NP_001026048.1 | 3 - 4 | 314 | 94 | |
XM_015296882.2 | XP_015152368.2 | 5 - 6 | 1122 | 94 | ||
XM_015296883.2 | XP_015152369.2 | 5 - 6 | 1071 | 94 | ||
XM_015296886.2 | XP_015152372.1 | 4 - 5 | 464 | 94 | ||
XM_015296887.2 | XP_015152373.1 | 4 - 5 | 292 | 94 | ||
XM_025142516.1 | XP_024998284.1 | 4 - 5 | 1212 | 94 | ||
XM_025142517.1 | XP_024998285.1 | 9 - 10 | 1571 | 94 | ||
XM_025142518.1 | XP_024998286.1 | 3 - 4 | 1015 | 94 | ||
GenBank mRNA | AJ720450.1 | - | 3 - 4 | 314 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |