Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030877.1 | NP_001026048.1 | 10 | 1334 | 135 | |
XM_015296882.2 | XP_015152368.2 | 12 | 2142 | 135 | ||
XM_015296883.2 | XP_015152369.2 | 12 | 2091 | 135 | ||
XM_015296884.2 | XP_015152370.2 | 11 | 2091 | 135 | ||
XM_015296886.2 | XP_015152372.1 | 11 | 1484 | 135 | ||
XM_015296887.2 | XP_015152373.1 | 11 | 1312 | 135 | ||
XM_025142516.1 | XP_024998284.1 | 11 | 2232 | 135 | ||
XM_025142517.1 | XP_024998285.1 | 16 | 2591 | 135 | ||
XM_025142518.1 | XP_024998286.1 | 10 | 2035 | 135 | ||
GenBank mRNA | AJ720450.1 | - | 10 | 1334 | 135 | |
AY332742.1 | - | 7 | 1161 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |