Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM46A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031085.1 | NP_001026256.1 | 1 - 2 | 816 | 114 | |
XM_015284627.2 | XP_015140113.2 | 3 - 4 | 688 | 114 | ||
XM_015284628.2 | XP_015140114.2 | 2 - 3 | 605 | 114 | ||
XM_015284629.2 | XP_015140115.2 | 2 - 3 | 604 | 114 | ||
GenBank mRNA | AJ720583.1 | - | 1 - 2 | 816 | 114 | |
CR390155.1 | - | 1 - 2 | 440 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |