Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031198.1 | NP_001026369.1 | 6 - 7 | 678 | 147 | |
XM_015287340.1 | XP_015142826.1 | 6 - 7 | 927 | 147 | ||
XM_015287341.2 | XP_015142827.1 | 6 - 7 | 851 | 147 | ||
GenBank mRNA | AJ720818.1 | - | 6 - 7 | 678 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |