Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031205.1 | NP_001026376.1 | 5 - 6 | 1484 | 82 | |
XM_015287393.2 | XP_015142879.1 | 5 - 6 | 1492 | 82 | ||
XM_015287394.2 | XP_015142880.1 | 5 - 6 | 1484 | 82 | ||
XM_015287395.2 | XP_015142881.1 | 5 - 6 | 1422 | 82 | ||
XM_025150750.1 | XP_025006518.1 | 8 - 9 | 1893 | 82 | ||
GenBank mRNA | AJ720508.1 | - | 5 - 6 | 1484 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |