Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031220.1 | NP_001026391.1 | 4 - 5 | 752 | 128 | |
XM_015288120.2 | XP_015143606.1 | 3 - 4 | 810 | 128 | ||
XM_015288121.2 | XP_015143607.1 | 3 - 4 | 810 | 128 | ||
XM_015288128.2 | XP_015143614.1 | 11 - 12 | 5069 | 128 | ||
GenBank mRNA | AJ720565.1 | - | 4 - 5 | 752 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |