Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031220.1 | NP_001026391.1 | 6 - 7 | 1069 | 149 | |
XM_015288120.2 | XP_015143606.1 | 5 - 6 | 1127 | 149 | ||
XM_015288128.2 | XP_015143614.1 | 13 - 14 | 5386 | 149 | ||
GenBank mRNA | AJ720565.1 | - | 6 - 7 | 1069 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |