Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031222.1 | NP_001026393.1 | 5 - 6 | 687 | 104 | |
XM_015288131.2 | XP_015143617.2 | 6 - 7 | 778 | 104 | ||
XM_015288132.2 | XP_015143618.2 | 5 - 6 | 573 | 104 | ||
XM_015288133.2 | XP_015143619.2 | 6 - 7 | 753 | 104 | ||
XM_015288134.2 | XP_015143620.2 | 6 - 7 | 969 | 104 | ||
XM_015288135.2 | XP_015143621.2 | 5 - 6 | 851 | 104 | ||
GenBank mRNA | AJ720344.1 | - | 5 - 6 | 687 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |