Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AvBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001193.1 | NP_001001193.1 | 3 - 4 | 265 | 72 | |
GenBank mRNA | AY621308.1 | - | 3 - 4 | 265 | 72 | |
DQ677637.1 | - | 2 - 3 | 210 | 72 | ||
DQ858303.1 | - | 2 - 3 | 210 | 72 | ||
DQ858316.1 | - | 2 - 3 | 210 | 72 | ||
DQ858329.1 | - | 2 - 3 | 210 | 72 | ||
DQ858343.1 | - | 2 - 3 | 210 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |