Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001461.1 | NP_001001461.1 | 1 - 2 | 374 | 74 | |
GenBank mRNA | AF019621.1 | - | 1 - 2 | 374 | 74 | |
AY448007.1 | - | 1 - 2 | 374 | 74 | ||
GU075927.1 | - | 1 - 2 | 374 | 74 | ||
GU075928.1 | - | 1 - 2 | 374 | 74 | ||
GU075929.1 | - | 1 - 2 | 374 | 74 | ||
KF721281.1 | - | 1 - 2 | 171 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |