Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001530.1 | NP_001001530.1 | 12 - 13 | 1769 | 84 | |
XM_015291385.2 | XP_015146871.1 | 12 - 13 | 84 | |||
GenBank mRNA | AY463529.1 | - | 12 - 13 | 1769 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |