Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001606.1 | NP_001001606.1 | 2 - 3 | 193 | 83 | |
GenBank mRNA | AY534899.1 | - | 2 - 3 | 193 | 83 | |
BX932651.1 | - | 2 - 3 | 271 | 83 | ||
BX934851.1 | - | 2 - 3 | 254 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |