Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001743.1 | NP_001001743.1 | 1 - 2 | 235 | 83 | |
GenBank mRNA | AF199610.1 | - | 1 - 2 | 235 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |