Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001745.1 | NP_001001745.1 | 1 - 2 | 76 | 74 | |
XM_025149542.1 | XP_025005310.1 | 1 - 2 | 243 | 74 | ||
XM_025149543.1 | XP_025005311.1 | 1 - 2 | 242 | 74 | ||
GenBank mRNA | Y14063.1 | - | 76 | 74 | ||
Y14064.1 | - | 56 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |