Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001748.2 | NP_001001748.1 | 1 - 2 | 191 | 66 | |
GenBank mRNA | BX935062.1 | - | 1 - 2 | 160 | 66 | |
M18539.1 | - | 1 - 2 | 168 | 66 | ||
M32293.1 | - | 1 - 2 | 192 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |