Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001751.2 | NP_001001751.1 | 10 - 11 | 1093 | 90 | |
GenBank mRNA | AJ250337.1 | - | 10 - 11 | 1094 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |