Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002242.1 | NP_001002242.1 | 6 - 7 | 561 | 141 | |
XM_015296419.2 | XP_015151905.1 | 6 - 7 | 616 | 141 | ||
XM_025142266.1 | XP_024998034.1 | 7 - 8 | 998 | 141 | ||
XM_025142267.1 | XP_024998035.1 | 6 - 7 | 554 | 141 | ||
XM_025142268.1 | XP_024998036.1 | 6 - 7 | 553 | 141 | ||
XM_025142269.1 | XP_024998037.1 | 6 - 7 | 553 | 141 | ||
XM_025142270.1 | XP_024998038.1 | 6 - 7 | 554 | 141 | ||
GenBank mRNA | M59456.1 | - | 6 - 7 | 561 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |