Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004369.1 | NP_001004369.1 | 7 - 8 | 1038 | 116 | |
XM_015278892.2 | XP_015134378.1 | 8 - 9 | 1179 | 116 | ||
XM_015278893.2 | XP_015134379.1 | 7 - 8 | 1265 | 116 | ||
GenBank mRNA | X62531.1 | - | 1038 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |