Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004378.2 | NP_001004378.1 | 2 - 3 | 387 | 116 | |
GenBank mRNA | AJ392770.1 | - | 2 - 3 | 387 | 116 | |
AY393848.1 | - | 2 - 3 | 285 | 116 | ||
BU137661.1 | - | 1 - 2 | 131 | 116 | ||
CR386189.1 | - | 2 - 3 | 367 | 116 | ||
M24193.1 | - | 2 - 3 | 367 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |