Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006415.1 | NP_001006415.1 | 4 - 5 | 793 | 121 | |
XM_025148781.1 | XP_025004549.1 | 4 - 5 | 830 | 121 | ||
GenBank mRNA | AJ719453.1 | - | 4 - 5 | 793 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |