Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321516.1 | NP_001308445.1 | 3 - 4 | 681 | 88 | |
GenBank mRNA | L00677.1 | - | 3 - 4 | 683 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |