Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321516.1 | NP_001308445.1 | 6 - 7 | 1328 | 96 | |
GenBank mRNA | BX935744.1 | - | 4 - 5 | 648 | 96 | |
L00677.1 | - | 6 - 7 | 1330 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |