Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2AF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204175.2 | NP_989506.1 | 2 - 3 | 248 | 115 | |
XM_015297931.2 | XP_015153417.1 | 2 - 3 | 658 | 115 | ||
GenBank mRNA | AB052869.1 | - | 2 - 3 | 206 | 115 | |
AJ720333.1 | - | 2 - 3 | 248 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |